Learn More
Molecular Probes™ Pacific Orange™ Protein Labeling Kit
From start to finish, it takes less than three hours to label a protein and purify the resulting conjugate using the Pacific Orange™ Protein Labeling Kit.
Brand: Molecular Probes™ P30016
Code : 50
Additional Details : Weight : 0.00450kg
Description
From start to finish, it takes less than three hours to label a protein and purify the resulting conjugate using the Pacific Orange™ Protein Labeling Kit. Although optimized for 1 mg of an IgG antibody, this kit can be used to make conjugates of other proteins larger than ∼40,000 daltons. The kit provides sufficient material to perform three protein conjugations and purifications. To label 100 μg of an IgG, we have the Pacific Orange™ Monoclonal Antibody Labeling Kit (Cat. No. P30014).
View a selection guide for all Protein Labeling Kits.
View the Fluorophore Selection Guide.
Specifications
Pacific Orange™ Protein Labeling Kit | |
1 Kit | |
Kit contains everything that is required to perform three separate labeling reactions and to purify the resulting conjugates. The Pacific Orange™ reactive dye has a succinimidyl ester moiety that reacts efficiently with primary amines of proteins | |
Store at 2–8°C. | |
Pacific Orange™ | |
405 |
Labeling chemistry, protein labeling and crosslinking | |
Pacific Orange™ Protein Labeling Kit | |
3 Reactions | |
Fluorescence | |
551 |