missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Support Stand

For Use With: Heracell 240i, single chamber

Brand:  Thermo Scientific™ 50065753

Code : 51

Additional Details : Weight : 0.80000kg

Product Code. 11811641

  • £940.00 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

For Use With: Heracell 240i, single chamber

Specifications

Specifications

Heracell 240i, single chamber
Support Stand
78 cm
LED
30.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers