missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Set of Three Heratrays

Material: Copper; For Use With: Heracell 240i

Brand:  Thermo Scientific™ 50065806

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 10462974

  • £330.43 / Pack of 3
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

Material: Copper; For Use With: Heracell 240i

Specifications

Specifications

Heracell 240i
Copper
0.33 in.
LED
Tray
0.84 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers