missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Polystyrene Roller Bottles

Catalog Number 11917904
Click to view available options
Cell Growth Area:
1450 cm2
850 cm2
Bottle Surface:
Pleated
Smooth
Smooth surface, deep indent
Smooth surface, shallow indent
Packaging:
2/Pk., 20/Case
2/Pk., 20/Case; Double-bagged
20/Pk., 20/Case
20/Pk., 20/Case; Double-bagged
This item is not returnable. View return policy
This item is not returnable. View return policy

Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.

Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6

  • One-piece seamless design: Reduces possibility of leaking through a seam
  • Shallow and deep indents: Unique ergonomic design makes the roller bottle excellent for both manual and automated handling
  • Proprietary manufacturing process: Greater impact resistance
  • Vertical expanded surfaces version: Vertical pleat orientation facilitates emptying and reduces retention of product
Recommended for:
Cell Culture Expansion, Vaccine Production, Antibody Production
TRUSTED_SUSTAINABILITY
Bottle Surface Smooth surface, shallow indent
Closure Material HDPE
Closure Type EZ Twist Non-vented Cap
Product Type Roller Bottle
Color Clear
For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production
Material Polystyrene
No. per Case 20
No. per Pack 20
Packaging 20/Pk., 20/Case; Double-bagged
Sterility Sterile
Cell Growth Area 850 cm2
Graduations Yes
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Round
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'