Learn More
Invitrogen™ Novex™ 12% Zymogram (Casein) Protein Gels, 1.0mm, 12 well
Excellent tools for detecting and characterizing proteases that utilize casein or gelatin as a substrate
Brand: Invitrogen™ EC64052BOX
Code : 50
Additional Details : CAS Number : 53123-88-9 Weight : 0.14000kg
Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
Description
- Proteases are run on a gel containing one of these substrates under denaturing conditions and visualized as clear bands against a dark background following a simple renaturing, developing, and staining protocol
- Zymogram gels are commonly used to detect matrix metalloproteinases
- Tris-Glycine gels with a substrate (gelatin or casein) incorporated into the gel
- Made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water and casein or gelatin as the substrate; All have a 4% stacking gel
Sample Loading Volume: 20μL; 1D Well
- The 12% Novex Zymogram (Casein) Gel is a 12% Tris-Glycine gel with 0.05% casein as the substrate
- Sensitivity Level: 12% Zymogram Casein Gel: 7 x 10-4 units of trypsin
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
12% | |
Zymogram Casein | |
Protein | |
12-well | |
1D Well |
1 mm | |
Homogeneous Page | |
20 to 120 kDa | |
10/Pk. |
For Research Use Only. Not for use in diagnostic procedures.