Learn More
Nitinol wire, , 0.58mmAlfa Aesar™
Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface
£78.40 - £885.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410940 SDS View Documents Alfa Aesar
044951.G6
10m This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11420940 SDS View Documents Alfa Aesar
044951.H2
50m This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11430940 SDS View Documents Alfa Aesar
044951.H5
150m This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»