Learn More
Thermo Scientific™ National Scientific Inserts for Target LoVial™ wide-opening vials
With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.
Brand: Thermo Scientific™ C4010-630TS
Code : 17
Additional Details : Weight : 0.05000kg
Description
With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.
- Pulled point inserts are an economical choice for noncritical applications
- Nominal O.D. of 6mm to fit Target wide-opening vials
For use with:
- Target LoVial Wide-Opening vials
- Snap-It™ Wide-Opening Snap Cap vials
- Target DP™ vials
- Target Wide-Opening vials
Specifications
150uL Glass Conical with Top Spring | |
Insert is suspended from the rim of the vial | |
Target LoVial Wide-Opening Vials |
150 μL | |
National |