missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot

Prepare large volume media and cultures with these round polycarbonate Clearboys. Lighter, safer and more durable than glass. Useful for visual quality inspections.

£128.00 - £165.00

Specifications

Diameter (Metric) Inner Neck 66 mm
Autoclavable Yes
Closure Size 83 B
Graduations Molded-in graduations in 1 gal. and 5 L increments
Material Polycarbonate
View More Specs

Products 2
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10722332 View accessory products View Documents Thermo Scientific™
2317-0020
10 L 83 B £128.00
Each View alternative products
Save Up to 
»
In Stock 8
Please to purchase this item. Need a web account? Register with us today!
10684071 View accessory products View Documents Thermo Scientific™
2317-0050
20 L 83 B £165.00
Each View alternative products
Save Up to 
»
In Stock 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Features:
  • Molded in graduations in 1 gal and 5L increments for ease in dispensing
  • Molded in spigot boss provides strength and leak resistance
  • Polycarbonate is glass-clear for easy visual inspection and volume measurements
  • Suitable for mixing, storing and freezing aqueous-based reagents and biologicals
  • Designed with 83mm neck to accept TopWorks and other Nalgene accessories for versatility
  • Graduated
  • Leakproof†
  • Autoclavable for sterile applications 

Includes:

Polypropylene spigot screw closure and TPE gasket. See autoclaving instructions for more information.

Note:

Completely disengage closure threads and disassemble spigot before autoclaving
Follow product insert instructions for autoclaving to ensure proper function
Note: Tubing adaptor is not autoclavable

Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com

Specifications

Specifications

66 mm
83 B
Polycarbonate
4
Yes
Molded-in graduations in 1 gal. and 5 L increments
Spigot
1

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.