Learn More
Miele Professional™ Mobile Injector, Module A 301/3
Module A 301/3 for the optimum loading of butyrometers
Brand:  Miele Professional™ 11116950
Code : NEW
Additional Details : Weight : 6.00000kg
Product Code. 15457704
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Equipped with 18 SD-B injector nozzles for butyrometers
  • Nozzle spacing 78 mm across the width, 84 mm across the depth