Learn More
Thermo Scientific™ Microscope Slides, Cut, Square
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
£15.90 - £1201.73
Specifications
Material Soda-lime glass
Color Extra White
View More Specs
Products 12
Product Code Brand Dimensions (L x W) Quantity Thickness (Metric) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Quantity Thickness (Metric) Price Quantity & Availability  
15388071 View Documents Thermo Scientific™
AAAC0240024002EB0MNZ#0#G#
24 x 24mm 100 pieces in paper 1mm £201.57
Pack of 1000 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
15642239 View Documents Thermo Scientific™
AAAC0220076002EB0MNZ#0#G#
76 x 22mm 100 pieces in paper 1mm £15.90
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
15917165 View Documents Thermo Scientific™
AAAC1000100002EB0MNZ#0#G#
100 x 100mm 100 pieces in paper 1mm £80.42
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
13487486 View Documents Thermo Scientific™
AAAC0590059002EB0MNZ#0#G#
59 x 59mm 100 pieces in paper 1mm £453.00
Pack of 1000 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
15103584 View Documents Thermo Scientific™
AAAC0260026002EB0MNZ#0#G#
26 x 26mm 100 pieces in paper 1mm £259.00
Pack of 1000 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
15353786 View Documents Thermo Scientific™
AAAC0680068002EB0MNZ#0#G#
68 x 68mm 100 pieces in paper 1mm £32.75
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
15513051 View Documents Thermo Scientific™
AAAC0250025003EB0MNZ#0#G#
25 x 25mm 100 pieces in paper 1 - 1.2mm £279.68
Pack of 1000 View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Add to basket
13489518 View Documents Thermo Scientific™
AAAC1000100002EB0MNZ#0#G#
100 x 100mm 100 pieces in paper 1mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12361533 View Documents Thermo Scientific™
AAAC0220076002EB0MNZ#0#G#
76 x 22mm 100 pieces in paper 1mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15310756 View Documents Thermo Scientific™
AAAC0500050007EA0MNZ#0#G#
50 x 50mm 50 pieces in paper 1.8 - 2.0mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15235707 View Documents Thermo Scientific™
AAAC0250025002BB0MNZ#0#G#
25 x 25mm 100 pieces in paper 1mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15522802 View Documents Thermo Scientific™
AAAC0250025000EB0MNZ#0#G#
25 x 25mm 100 pieces in paper 0.6 - 0.8mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
Soda-lime glass
Extra White