Learn More
Lovibond™ Powder Pack™ Reagents: Iron, Total; TPTZ ; 10mL
Pre-measured, unit dose reagent for water quaility testing
Brand: Lovibond™ 530563
Code : NEW
Additional Details : Weight : 0.30000kg
Description
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open the package
- Easily create a “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in the corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
US EPA | |
10 mL | |
Iron colorimeter, colorimeter, spectrophotometer | |
1,10-Phenanthroline | |
0.33 oz. |
Sodium Dithionite 30 to 40%, Disodium Disulphite 20 to 30%, Sodium Thiosulphate 30 to 40%, 1, 10-phenanthroline monohydrochloride monohydrate 1 to 5%, Sodium Citrate Tribasic Dihydrate 1 to 10% | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
0 to 3mg/L | |
Iron Ferro Reagent |
Safety and Handling
- X1000 REACTIF poudre VARIO FER 10ML
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium dithionite, disodium disulphate,sodium carbonate, 1,10-phenanthroline, methenamine