Learn More
Thermo Scientific Sorvall Furnace, Box; Thermo Scientific Lindberg/Blue M Moldatherm; 100 to 1100C; 18.4 L; Digital/OTP Control; 240V, 50/60Hz; 3500W
Brand:  Thermo Scientific Sorvall BF51794C-1
Furnace, Box; Thermo Scientific Lindberg/Blue M Moldatherm; 100 to 1100C; 18.4 L; Digital/OTP Control; 240V, 50/60Hz; 3500W
Code : Z4
Additional Details : Weight : 59.00000kg
Product Code. 15206229