missing translation for 'onlineSavingsMsg'
Learn More

LEC Refrigeration Refrigerator PE109 pharmacy, bench top, 470mm x 465mm x 530mm (w x d x h), 37L

Brand:  LEC Refrigeration PE109C

Refrigerator PE109 pharmacy, bench top, 470mm x 465mm x 530mm (w x d x h), 37L

Code : 03

Additional Details : Weight : 20.00000kg

Product Code. 12998674

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.