missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BPC Bag with 3 Ports, MPC Insert, MPC Body, and Luer Lock

Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

£29.19 - £1010.00

Specifications

Certifications/Compliance cGMP
Description Two-dimensional disposable cell culture/bioprocess bag
Film Description CX5-14 Single Web Film
Port Detail 3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in. Female Luer Lock
Sterilization Method Gamma Irradiated
View More Specs

Products 10
Product Code Brand Capacity (Metric) Quantity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Quantity Price Quantity & Availability  
15556849 View Documents Thermo Scientific™
SH30712.01
5L 1 £955.00
Pack of 20 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15566849 View Documents Thermo Scientific™
SH30712.02
10L 1 £985.00
Pack of 20 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15576849 View Documents Thermo Scientific™
SH30712.03
20L 1 £830.00
Pack of 16 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15586849 View Documents Thermo Scientific™
SH30712.04
50L 1 £1010.00
Pack of 12 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15596849 View Documents Thermo Scientific™
SH30712.05
2L 1 £830.00
Pack of 20 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10004733 View Documents Thermo Scientific™
SH30712.02
10L 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10298183 View Documents Thermo Scientific™
SH30712.04
50L 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10465235 View Documents Thermo Scientific™
SH30712.01
5L 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10552974 View Documents Thermo Scientific™
SH30712.05
2L 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10414285 View Documents Thermo Scientific™
SH30712.03
20L 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

cGMP
CX5-14 Single Web Film
Gamma Irradiated
No
Two-dimensional disposable cell culture/bioprocess bag
3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in. Female Luer Lock
Ambient