missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BPC Bag with 3 Ports, Luer Lock

Simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

£31.70 - £1545.00

Specifications

Certifications/Compliance cGMP
Description Two-dimensional disposable cell culture/bioprocess bag
Film Description CX5-14 Single Web Film
Port Detail Female 1/8” Luer Connector, Male 3/8” MPC Connector, Female 3/8” MPC Connector
Sterilization Method Gamma Irradiated
View More Specs

Products 10
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15506859 View Documents Thermo Scientific™
SH30713.01
5L £1545.00
Pack of 34 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15513983 View Documents Thermo Scientific™
SH30713.02
10L £1150.00
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15516859 View Documents Thermo Scientific™
SH30713.03
20L £1230.00
Pack of 25 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15526859 View Documents Thermo Scientific™
SH30713.04
50L £1220.00
Pack of 15 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15536859 View Documents Thermo Scientific™
SH30713.05
2L £955.00
Pack of 25 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10731694 View Documents Thermo Scientific™
SH30713.01
5L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10677584 View Documents Thermo Scientific™
SH30713.04
50L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10389743 View Documents Thermo Scientific™
SH30713.02
10L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10414475 View Documents Thermo Scientific™
SH30713.05
2L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10453485 View Documents Thermo Scientific™
SH30713.03
20L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

cGMP
CX5-14 Single Web Film
Gamma Irradiated
No
Two-dimensional disposable cell culture/bioprocess bag
Female 1/8” Luer Connector, Male 3/8” MPC Connector, Female 3/8” MPC Connector
Ambient