missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BPC Bag with 2 Ports, Luer Lock and MPC Insert

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Brand:  Thermo Scientific™ SH30658.13

Code : 84

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11516311

  • £186.00 / Pack of 10

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport &

    Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

Two-dimensional disposable cell culture/bioprocess bag; SH30658.13
Female Luer Lock, 12in. of 1/4in. Tubing; Male Luer Lock, 12in. of 1/4in. Tubing
Ambient
250mL
CX5-14 Single Web Film
Gamma Irradiated
12 in. of 1/4 in. Tubing
Videos
Documents