missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Protector™ Standard Storage Cabinet
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
POA
Specifications
Material | Epoxy-Coated Steel |
---|---|
Product Type | Storage Cabinet |
For Use With (Application) | Ideal for storage and venting of small laboratory instrument and mild acid and other reagent |
Includes | Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. |
Load Capacity (Metric) Centralized | 362.8kg |
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Order Info
Work surface and interior shelf kits are sold separately
Specifications
Epoxy-Coated Steel | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
362.8kg |
Storage Cabinet | |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. |