Learn More
Labconco™ Activated Carbon Solvent Trap for Rotary Vane Vacuum Pumps
Activated Carbon Solvent Trap protects a rotary vane vacuum pump from organic solvent vapors.
Brand:  Labconco™ 7772500
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 10535315
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Secondary trap connects between the vacuum pump and the freeze dryer or evaporator to provent migration of organic solvents into the pump interior. Cartridge contains 11 ounces of activated carbon media. 3/4 in. OD inlet and outlet connections. Includes 20 in. rubber tubing, 3/4 in. ID x 3/8 in. wall, and one clamp.
Specifications
Specifications
Rotary vane vacuum pumps
20 in. rubber tubing, 0.75 I.D. x 0.375 in. wall and one clamp
Activated Carbon Solvent Trap