Learn More
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Calibrated “To Contain”
POA
Description
Calibrated “To Contain”
- Permanently marked with individual serial number
- Individually calibrated and certified against equipment whose calibration is traceable to NIST™
- Cylindrical body for better mixing and draining
- Easier to withdraw samples using pipets
- Wide base provides stability
- Marking spot
- Ground glass stopper
- 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
Compliance
Designed from requirements of ASTM™ E 237, Class A serialized.