Learn More
Invitrogen™ PureLink™ Genomic Elution Buffer
Allows highest DNA binding of any column method
Brand: Invitrogen™ K182305
Code : 50
Additional Details : Weight : 0.44000kg
Includes: PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.
Description
PureLink Genomic DNA Mini Kit and PureLink 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format. For higher throughput, use PureLink 96 Genomic DNA Kit. In this 96-well version, sample processing is performed with either vacuum manifold or centrifugation at only 2,250g, allowing you more flexibility in centrifuge choice.
- Obtain higher gDNA yields and purity - optimized spin-column membrane and buffer formulations
- Save time and money - use with a Large range of sample sizes (e.g., 100μL to 1mL blood); 96-well version does not require special equipment
- Increase flexibility - one kit works with a variety of sample types
- PureLink Genomic DNA Mini Kit protocol improves upon the easy, familiar spin-column methods currently in use
- Columns are newly configured to bind and release DNA more efficiently
- Samples are lysed in optimized buffer formulated to enhance proteinase K activity and eliminate protein contamination
- Powerful wash buffers remove all traces of protein and salt
- DNA is eluted in low-salt buffer to allow for pH stabilization of DNA in storage
- Semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations
- With the PureLink Genomic DNA Kits, get greater yields of higher-purity gDNA (determined by A260/A280 measurements) with all of your sample types - from bacteria to tissues to blood to cells
- The PureLink 96 Genomic DNA Kit outperforms competitor products in both DNA yield and purity for multiple sample types, including blood
- A260/A280 and A260/A230 ratios of PureLink gDNA are higher than those obtained with other kits, indicating no contamination carryover of protein or guanidine using enhanced PureLinkLysis and Wash Buffers
Automated Genomic DNA Purification, Automated Nucleic Acid Purification, DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Animal Tissues & Cells, gDNA from Bacteria, gDNA from Blood (Medium-Scale, 100 μl-1mL)
Order Info
Shipping Condition: Room Temperature
Specifications
Elution Buffer | |
160mL |
For Research Use Only. Not for use in diagnostic procedures.