Learn More
Liebherr™ Water Filtration Cartridges for Ice Machines
Remove water hardness impurities prior to making ice flakes. Liebherr™ Water Filtration Cartridges for Ice Machines works as a de-scaler, de-carbonizer and de-ironizer, prevention of algae and corrosion, deodorizer and water filter.
POA
Specifications
Type | Anticalculus Flocculator Double Cartridge |
---|---|
For Use With (Equipment) | Ice machines |
Description
Liebherr™ Ice Makers create granular wet ice flakes for rapid cooling. Units with air or water condensers are available (water condensers are recommended for hot environments).
- The CACF1 water filtration device is essential for removing impurities from the water
- The complete device includes two in-line holders with 1 cartridge each
- Flocculator cartridge: transforms calcium carbonates into stable, suspended calcium salts that cannot clump together
- Filter cartridge: sifts the salts and retains impurities suspended in the water
- The treatment capacity of the CACF1 unit varies, depending on the hardness of the water
- Replace the cartridges at least 1x/year or when the pressure between in the inlet and the outlet of the unit drops by ≥500g
- Replacement cartridges are available separately (mfr. no. KIT2CART)
Specifications
Anticalculus Flocculator Double Cartridge | |
Ice machines |