Learn More
Invitrogen™ mirVana™ miRNA Inhibitor, Negative Control #1
mirVana™ miRNA Inhibitor, Negative Control #1
Brand:  Invitrogen™ 4464079
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15541511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
We have now released the second generation of miRNA inhibitors,mirVana™ miRNA Inhibitors which:
  • Display better potency than lead competitor's product
  • Offer maximum and consistent effect in vitro at low miRNA inhibitor concentration
  • Are non toxic
  • Target the most updated Sanger content
  • Can be used in vitro and in vivo, offering consistency throughout your entire research project
    Positive and negative controls are also available for use along with mirVana™ miRNA Inhibitors in miRNA down-regulation experiments.
    mirVana™ miRNA Inhibitor Negative Control #1
    mirVana™ miRNA Inhibitor Negative Control #1 is intended for use as a negative control for experiments using mirVana™ miRNA Inhibitors. Transfect this negative control using the same methodology as for your positive control (such as let-7c Positive Control) and experimental mirVana™ miRNA Inhibitors. Use target expression from negative control-transfected samples as a baseline for evaluation of the effect of the control and experimental miRNA inhibitor on target gene expression.
    First Generation Inhibitor Controls
    Our first-generation miRNA inhibitors (and controls), Invitrogen™ Anti-miR™ miRNA Inhibitors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Inhibitors. These first-generation inhibitors are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Inhibitors and their corresponding controls.
Specifications
Specifications
-20°C
miRNA and non-coding RNA analysis
mirVana™ Negative Control #1 miRNA Inhibitor
250 nmol
mirVana™ miRNA Inhibitor, 250nmol, Unconjugated Dye, Anti-miRNA RNAi Type, Negative Control Type, Used for miRNA and Non-Coding RNA Analysis, HPLC Purification, –20°C Storage Temperature, For Research Use Only
Unconjugated
HPLC purified