missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, 48%, AR, For ACS analysis, Macron Fine Chemicals™
612.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Specifications
Color | Colorless |
CAS | 7664-39-3 |
Assay Percent Range | 48.75 to 49.2 % |
Quantity | 2.5 L |
Molecular Formula | FH |
Physical Form | Corrosive Fuming Solution |
MDL Number | MFCD00011346 |
Synonym | hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
SMILES | [H+].[F-] |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'