Learn More
HyClone™ Fetal Bovine Serum (U.S.), Standard
Highly filtered and meets virus testing requirements.
Brand: HyClone products Cytiva SH30088.03IH
Code : 85
Additional Details : Weight : 0.01000kg
Description
Serum Collection:
- From collection to final packaging
- Every step is documented
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Aseptically packaged in a Class 100 cleanroom
- Each lot of serum is tested to ensure quality and sterility
- Pooled after filtration and before final packaging
- Consistent quality from the first bottle filled to the last
- Increases consistency and minimizes variability with a lot of serum
Certifications
Manufactured under cGMP and ISO 9001 certified facilities.
FOR FURTHER MANUFACTURING OR RESEARCH USE. NOT FOR DIAGNOSTIC OR THERAPEUTIC USE.