Learn More
Invitrogen™ SCF Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHKITLG
362.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00400kg
Description
The Human Stem Cell Factor/KITLG (Hu SCF) ELISA quantitates Hu SCF in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu SCF. Principle of the method The Human SCF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.Specifications
P21583 | |
2 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
4254 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Immunology | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.74-2000 pg/mL | |
2pg/mL to 2000pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL- 12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
DCUA,DFNA69,FPH2,FPHH,KL-1,Kitl,MGF,SCF,SF,SHEP7 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
FPH2, KL-1, Kitl, MGF, SCF, SF, SHEP7 | |
1 hr. 20 min. |