Learn More
Thermo Scientific™ HEPA Filter for Sorvall™ LYNX 6000 Superspeed Centrifuge
Expand biosafety levels by using this optional HEPA Filter for Thermo Scientific™ Sorvall™ LYNX 6000 Superspeed Centrifuge.
Brand:  Thermo Scientific™ 75000011
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 15682899
  • £2505.00 / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket
Description
Description
Expand biosafety levels by using this optional HEPA Filter for Thermo Scientific™ Sorvall™ LYNX 6000 Superspeed Centrifuge.
Specifications
Specifications
HEPA Filter
Thermo Scientific Sorvall LYNX 6000 Superspeed Centrifuge
HEPA Filter