Learn More
Thermo Scientific™ HCT 116 (+hIGF-1) Control Lysate (1mg/mL)
Thermo Scientific™ HCT 116 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the RAS signaling pathway.
Brand: Thermo Scientific™ A37961
Additional Details : Weight : 0.01000kg
Description
- Validated for the expression RAS by western blotting and of NRAS, KRAS, and HRAS by mass spectrometry analysis
- Optimized - stimulation and lysis conditions optimized for detection of RAS signaling targets
- Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
- Stable for two years at -80°C
Specifications
Mass Spectrometry | |
Dry Ice | |
Frozen Liquid | |
0.5 mg (1 mg/mL) | |
Protein |
Mass Spectrometer | |
Sample Enrichment | |
Thermo Scientific™ | |
Control Lysate | |
Cell Lysate |
For Research Use Only. Not for use in diagnostic procedures.