missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ γ-Globulin from Bovine Plasma
Purity: 98%
POA
Specifications
Concentration | 25mg/mL |
---|---|
For Use With (Application) | Cell Biology and Analysis, Genomics, Molecular Biology |
Formulation | The product is lyophilized, in normal Saline solution (25mg/mL, 0.9%NaCl) |
Molecular Weight (g/mol) | ∽150000 |
Name | g-Globulin from Bovine Plasma |
Description
White, crystalline, lyophilized powder.
Specifications
25mg/mL | |
The product is lyophilized, in normal Saline solution (25mg/mL, 0.9%NaCl) | |
g-Globulin from Bovine Plasma | |
Bovine Plasma | |
Lyophilized |
Cell Biology and Analysis, Genomics, Molecular Biology | |
∽150000 | |
6 to 8 | |
0°C |