Learn More
Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. These gas guards are factory installed and built in to specified CO2 incubators.
£429.00
Products 1
Product Code Brand Description For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Description For Use With (Equipment) Price Quantity & Availability  
12101241 View Documents Thermo Scientific™
1900000
Gas Guard, CO2 £429.00
Each View alternative products
Save Up to 
»
Estimated Shipment
20-03-2020
Add to basket
Description
Description
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low