missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUSIP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562846
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DRKTPNQMKA KEGRNVYSSS RYDDYDRYRR SRSR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
FUSIP1 Polyclonal antibody specifically detects FUSIP1 in Human samples. It is validated for Immunohistochemistry.Specifications
FUSIP1 | |
Polyclonal | |
Unconjugated | |
SRSF10 | |
40 kDa SR-repressor protein; FUS interacting protein (serine/arginine-rich) 1; FUS interacting protein (serine-arginine rich) 1; Fus-associated protein with serine-arginine repeats; FUS-interacting protein (serine-arginine rich) 2; FUS-interacting serine-arginine-rich protein 1; Fusip1; FUSIP2; Neural-salient serine/arginine-rich protein; neural-salient SR protein; neural-specific SR protein; Nssr; NSSR1; NSSR2; PPP1R149; protein phosphatase 1, regulatory subunit 149; serine and arginine rich splicing factor 10; serine/arginine-rich splicing factor 10; serine/arginine-rich splicing factor 13A; serine-arginine repressor protein (40 kDa); SFRS13; SFRS13A; Splicing factor SRp38; splicing factor, arginine/serine-rich 13; splicing factor, arginine/serine-rich 13A; SR splicing factor 10; SRp38; SRrp40; Srsf10; Srsf13a; TASR; TASR1; TASR2; TLS-associated protein TASR; TLS-associated protein with Ser-Arg repeats; TLS-associated protein with SR repeats; TLS-associated serine-arginine protein; T | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10772 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75494 | |
SRSF10 | |
Recombinant protein corresponding to Human FUSIP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only