Learn More
FTSJD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556306
Detalles adicionales : Weight : 0.02000kg
Descripción
Immunogen sequence: GKPLVKDREA ELLYFADVCA GPGGFSEYVL WRKKWHAKGF GMTLKGPNDF KLEDFYSASS ELFEPYYGEG GIDGDGDITR PENISAFRNF VL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
FTSJD2 Polyclonal antibody specifically detects FTSJD2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
FTSJD2 | |
Polyclonal | |
Unconjugated | |
CMTR1 | |
1300018I05Rik; AI451264; Cap methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; cmtr1; FtsJ methyltransferase domain containing 2; ftsJ methyltransferase domain-containing protein 2; ftsjd2; hMTr1; hypothetical protein LOC393102; interferon-stimulated gene 95 kDa protein; ISG95; KIAA0082; LOW QUALITY PROTEIN: cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; LRRG00129; mKIAA0082; MTr1; RGD1307801; S-adenosyl-L-methionine-dependent methyltransferase ftsjd2; zgc:55336 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23070 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N1G2 | |
CMTR1 | |
Recombinant protein corresponding to Human FTSJD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only