missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562035
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VRRLQRAVAA VAATGPASAP GPGGGRSGPD RIALQPSGSL L Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
The protein encoded by this intronless gene belongs to the GSK-3-binding protein family. Studies show that this protein plays a role as a positive regulator of the WNT signaling pathway. It may be upregulated in tumor progression.Specifications
FRAT2 | |
Polyclonal | |
Unconjugated | |
FRAT2 | |
FRAT2; FRAT-2; frequently rearranged in advanced T cell lymphomas 2; frequently rearranged in advanced T-cell lymphomas 2; GSK-3; GSK-3 binding protein FRAT2; GSK-3-binding protein FRAT2; MGC10562; RP11-452K12.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75474 | |
FRAT2 | |
Recombinant protein corresponding to Human FRAT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only