Learn More
FOXN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551943
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LSLNKCFQKV ERSHGKVNGK GSLWCVDPEY KPNLIQALKK QPFSSASSQN GSLSPHYLSS VIKQNQVRNL KESDIDAAAA MMLLNTSIEQ GILECEKPLP LKTALQKKRS YGNAFHHPSA VRLQESDSLA TSIDPKEDHN YSA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 84%.
FOXN2 Polyclonal antibody specifically detects FOXN2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
FOXN2 | |
Polyclonal | |
Unconjugated | |
FOXN2 | |
3230402J05Rik; 6030465J18Rik; Fkh19; forkhead box N1; forkhead box N2; forkhead box protein N2; forkhead homolog 19; Foxn1; FOXN2; HTLF; Human T-cell leukemia virus enhancer factor; RGD1562291 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3344 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P32314 | |
FOXN2 | |
Recombinant protein corresponding to Human FOXN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only