missing translation for 'onlineSavingsMsg'
Learn More

Sodium formate, 99%, Thermo Scientific Chemicals

Catalog Number p-3757418
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
141-53-7
CHNaO2
68.007
MFCD00013101
HLBBKKJFGFRGMU-UHFFFAOYSA-M
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
2723810
CHEBI:62965
sodium;formate
C(=O)[O-].[Na+]
141-53-7
CHNaO2
68.007
MFCD00013101
HLBBKKJFGFRGMU-UHFFFAOYSA-M
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
2723810
CHEBI:62965
sodium;formate
C(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 258.0°C to 261.0°C
Color White
Boiling Point 360.0°C
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula HCO2Na
Quantity 1 kg
Beilstein 02, 8
Fieser 8458
Merck Index 15, 8756
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, insoluble in ether
Formula Weight 68.01
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Sodium formate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'