missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ FISH Tag™ DNA Green Kit, with Alexa Fluor™ 488 dye
FISH Tag™ DNA Green Kit, with Alexa Fluor™ 488 dye
Brand: Molecular Probes™ F32947
Code : 50
Additional Details : Weight : 0.00450kg
Description
FISH Tag™ DNA Kits provide a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. Choose from FISH Tag™ DNA Kits in three Alexa Fluor™ colors or get the FISH Tag™ DNA Multicolor Kit, which contains four spectrally distinct dyes to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content.FISH Tag™ Kit Specifications:
Complete Workflow Solution for FISH
FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.
More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence | |
Alexa Fluor 488 | |
DNA In Situ Hybridization Assay Kit | |
Not Target-Specific |
Green | |
Kit | |
Alexa Fluor™, FISH Tag™ | |
DNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.