missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Filter
Brand: Cytiva 10549778
Code : A0
Additional Details : Weight : 0.15000kg
Specifications
150 x 165 mm | |
150 x 165mm |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Flammable solid Category 1