Learn More
Thermo Scientific™ Maxima™ Hot Start Green 2X PCR Master Mix
Save time preparing samples with this ready-to-use 2x hot start master mix containing tracking dyes and a density reagent for direct loading on gels after PCR.
£79.95 - £336.00
Specifications
Product Type | Fermentas™ Maxima™ Hot Start |
---|---|
For Use With (Application) | Standard PCR, RT-PCR |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
11893923
|
Thermo Scientific™
K1061 |
100 reactions of 50 µl |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11803933
|
Thermo Scientific™
K1062 |
500 reactions of 50 µl |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Maxima Hot Start Green PCR Master Mix (2X) is a ready-to-use solution containing Maxima Hot Start Taq DNA Polymerase, optimized hot start PCR buffer, Mg2+, and dNTPs. The master mix is supplemented with two tracking dyes and a density reagent that allows for direct loading of PCR products on a gel.
The dyes in the master mix do not interfere with PCR performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion. The master mix retains all features of Maxima Hot Start Taq DNA Polymerase. It is capable of amplification of up to 3 kb from genomic DNA.
Highlights
- Convenient - Maxima Hot Start Taq DNA Polymerase in a ready-to-use mix
- Hot start PCR - reaction set-up at room temperature
- Four minute activation time
- Direct loading of PCR product on gels
- High specificity and sensitivity of PCR
Specifications
Fermentas™ Maxima™ Hot Start | |
Standard PCR, RT-PCR |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.