Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
3M Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit is used with 3M™ Versaflo™ TR-600 series air purifying device.
£237.00 - £1350.00
Specifications
For Use With (Equipment) 3M Versaflo Powered Air Turbo TR-600 Series
View More Specs
Products 4
Product Code Brand Model Product Type Includes Price Quantity & Availability  
Product Code Brand Model Product Type Includes Price Quantity & Availability  
15509265
View Documents Promotion Details
3M™
7000144504
TR-641E Single Station Battery Charger Kit TR-640 Cradle, Power Supply and Lead
£237.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
15519265
View Documents Promotion Details
3M™
7000144505
TR-644E Four Station Battery Charger Kit Four TR-640 Cradles, a Baseplate, Power Supply and Lead
£1045.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
16100841
View Documents Promotion Details
3M™
7100020282
TR-644UK Four Station Battery Charger Kit
£1350.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
15710058
View Documents Promotion Details
3M™
7100021098
TR-641UK Single Station Battery Charger Kit TR-640 Cradle, Power Supply and Lead This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
  • Batteries may be stored on the charger, fully charged, ready for immediate use
Specifications
Specifications
3M Versaflo Powered Air Turbo TR-600 Series