Learn More
Merck™ Omnipore™ Membrane Filter
Compatible with virtually all solvents, acids and alkaline solutions
£209.00 - £347.00
Specifications
Color White
Material Polytetrafluoroethylene (PTFE)
Quantity (No. per Case) 100/pk.
Wettability Hydrophilic
View More Specs
Products 6
Product Code Brand Diameter (Metric) Outer Pore Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Pore Size Price Quantity & Availability  
10704784 View Documents Merck™
JGWP04700
47mm 0.2μm £347.00
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
23-12-2019
10775534 View Documents Merck™
JGWP09025
90mm 0.2μm £229.00
Pack of 25 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
10678453 View Documents Merck™
JGWP02500
25mm 0.2μm £209.00
Pack of 100 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
10706694 View Documents Merck™
JHWP02500
25mm 0.45μm £236.00
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
10-01-2020
10347703 View Documents Merck™
JHWP09025
90mm 0.45μm £229.00
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10501884 View Documents Merck™
JHWP04700
47mm 0.45μm £258.00
Pack of 100 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Description
Description
  • White, plain (non-gridded) filter surface
  • 65μm thick, 80% porosity
  • Hydrophilic PTFE membrane bonded to a high-density polyethylene support
  • Maximum operating temperature of 130°C
Specifications
Specifications
White
Polytetrafluoroethylene (PTFE)
100/pk.
Hydrophilic