missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane
Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays
£501.00
Specifications
Certifications/Compliance | ANSI, SBS |
---|---|
Format | Standard |
Membrane | Negatively Charged Phosphocellulose |
Array | 8 x 12 |
Pore Size | 0.65 μm |
Product Code | Brand | Product Type | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | No. per Pack | Price | Quantity & Availability | ||||
10245703
|
Merck
MSPHN6B10 |
MultiScreenHTS-PH Plate | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Two-piece plate design eliminates surface contact with individual well “drip directors,” prevents crosstalk between wells, allows membrane access, and provides easy, contamination-free stacking and improved seal with vacuum manifolds
- Extended skirt enables stacking directly with collection plates, protects recessed underdrain, provides ample surface for barcode labels, and improves vacuum seal
- Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
- Meet ANSI™/SBS 2004 plate specifications
- Standardized dimensions allow full compatibility with automated gripper arms, stackers, barcode labels, plate washers, and plate readers; aligned, rigid sidewalls permit easy handling by robotic gripper arms
- Opaque plastic to prevents light transfer between wells during direct scintillation counting
Specifications
ANSI, SBS | |
Negatively Charged Phosphocellulose | |
0.65 μm | |
MultiScreenHTS-PH Plate | |
0.05 to 0.25 mL | |
Non-sterile | |
0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44 cm) | |
8.55 cm |
Standard | |
8 x 12 | |
Opaque | |
Barex | |
96 | |
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge | |
1.28 cm | |
1.44 cm |