Learn More
Merck MF-Millipore™ Membrane Filters
Designed to provide strength, flexibility and broad chemical compatibility for critical biological separations
Brand: Merck AAWG0250C
19.84 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Code : 72
Additional Details : CAS Number : 9004-70-0 Weight : 0.07740kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
Not Autoclavable | |
MCE | |
Non-sterile | |
For analytical and research applications. | |
100/Pk. | |
Hydrophilic | |
25 mm |
White | |
0.8 μm | |
190 mL/min x cm2 | |
100Pack | |
Filter Membranes | |
0.98 in. | |
25 mm |
Safety and Handling
- .8 MCE WH GRD Asbestos Monitoring 100Pk
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use to extinguish.