Learn More
Merck Millipore Durapore™ Membrane Filter
Strength, flexibility, and broad chemical compatibility for critical biological separations
£257.67
Specifications
Color White
View More Specs
Products 1
Product Code Brand Diameter (Metric) Outside For Use With (Application) Material Pore Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outside For Use With (Application) Material Pore Size Price Quantity & Availability  
10339750
View Documents
Merck Millipore
SVLP09050
90mm For clarifying filtration of biological solution; particle monitoring Polyvinylidene Fluoride (PVDF) 5.0μm £257.67
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Add to basket
Description
Description
  • Protein binding to hydrophilic Durapore membranes is two orders of magnitude lower than nitro-cellulose or PTFE
Specifications
Specifications
White