missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ EGFR Recombinant Human Protein (w/o Catalytic Activity Domain), hIgG1-Fc Tag

Suitable for use in studying protein-protein interactions, in cell biology research applications, and as a protein standard

Brand:  Sino Biological™ 10001H02H250

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13402899

  • £865.00 / 250µg

Estimated Shipment date
from Supplier 02-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 645) of human EGFR (NP_005219) fused to Fc region of human IgG1 at C-terminus
Specifications

Specifications

Studying protein-protein interactions and cell biology research
1956
≥85% by SDS-PAGE
Human
Truncated, Recombinant
Epidermal growth factor receptor
Assay Standard
Mammalian
IgG1 Fc Tag
Lyophilized
95kd
5 x 50μg
−20° to −70°C
<1 EU/μg
EGFR
Growth Factors and Receptors
EGFR Recombinant Human Protein
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.