Learn More
DyNAmo Flash SYBR™ DyNAmo Flash SYBR Green qPCR Kit
Developed for fast real-time qPCR, the kit provides qPCR results faster than most SYBR Green kits without compromising the qPCR performance.
£116.00 - £2310.00
Specifications
Detection Method | SYBR |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Tbr DNA polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10379167
|
Thermo Scientific™
F415L |
500 reactions |
£514.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10334009
|
Thermo Scientific™
F415XL |
2500 Reactions |
£2310.00
1000 reactions |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10751120
|
Thermo Scientific™
F415S |
100 reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo Flash SYBR Green qPCR Kit gives reliable quantification and early Cq values. Due to the high signal intensity, DyNAmo Flash SYBR Green qPCR provides an excellent signal-to-noise ratio.
- Extremely fast protocols: Combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- Included dUTP allows the use of UNG for prevention of carry-over contamination
- Convenient 2X master mix and optimized protocol
Specifications
SYBR | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler | |
DyNAmo Flash | |
Gene Expression | |
DNA, dsDNA |
Tube | |
qPCR | |
Fast | |
DyNAmo Flash SYBR Green qPCR Kit | |
Real Time PCR SYBR Master Mix | |
2X (DyNAmo Flash SYBR Green qPCR Mix), 50X (ROX Dye) |
For Research Use Only. Not for use in diagnostic procedures.