missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioxane solution EU Pharmacopoeia, Fisher chemical™
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
Purity Grade Notes:
Eu Ph Ref: 1032001
Eu Ph Ref: 1032002
Specifications
Packaging | Bottle |
Quantity | 100 mL |
Type | Mass Spectrometry Standards |
Grade | Eu Ph Ref: 1032002 |
Chemical Name or Material | Dioxan stock solution. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'