missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chart Recorders and Paper

For Use With: Environmental Chamber (3962)

Brand:  Thermo Scientific™ 201156

Code : NEW

Additional Details : Weight : 2.70000kg

Product Code. 15593761

  • £2392.16 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.

Specifications

Specifications

Single Pen, 230V, 50/60 Hz, 6 in. 7 Day Chart Recorders; 0°C to 60°C (32°F to 140°F)
LED
Accessory, Chart Recorders and Paper
Environmental Chamber (3962)
0°C to 60°C
Customer Installed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers