Learn More
Thermo Scientific™ Data Output, 4 to 20 milliamp
For Use With: Environmental Chamber (3962)
Brand: Thermo Scientific™ 190512
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
4-20 milliamp Data Output | |
LED | |
Factory Installed |
Environmental Chamber (3962) | |
Data Output |