missing translation for 'onlineSavingsMsg'
Learn More

Corning™ LSE Digital Dry Bath Heater Blocks

£82.41 - £212.04

Specifications

For Use With (Equipment) LSE Digital Dry Bath Heater
View More Specs

Products 4
Product Code Brand Type For Use With (Application) Price Quantity & Availability  
Product Code Brand Type For Use With (Application) Price Quantity & Availability  
11363825 View Documents Corning™
480126
Dry Bath Block (Single) Holds 12 x 17mm tubes £82.41
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
11373825 View Documents Corning™
480127
Dry Bath Block (Single) Holds 6 x 20mm tubes £82.41
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
11333835 View Documents Corning™
480133
Tube Block (Dual) 384-well PCR plate £212.04
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
11313835 View Documents Corning™
480131
Tube Block (Dual) 96-well plate or 4 slides £198.16
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A variety of blocks are available for different sample containers.
Specifications

Specifications

LSE Digital Dry Bath Heater