Learn More
IBA Lifesciences Competent E. coli TOP10
Competent E. coli TOP10 are chemically competent cells for plasmid propagation
Brand: IBA Lifesciences 5-1600-020
Code : LE
Additional Details : Weight : 0.00900kg
Alert:
The purchase of this product requires a valid GMO end-user declaration.
Description
- Competent E. coli TOP10 cells are ready for heat shock transformation with vector DNA and its subsequent propagation.
- The chemically competent cells are provided as 20 separate one-shot reactions. Transformed cells can be selected by blue/white screening.
- The genotype of Competent E. coli TOP10 cells is F- mcrA (mrr-hsdRMS-mcrBC) 80lacZ M15 lacX74 recA1 ara 139 (ara-leu)7697 galU galK rpsL (StrR) endA1 nupG.
This product requires dry ice shipment and is shipped directly from our Headquarters in Germany. Please note, that it cannot be shipped from our US Distribution Center in Olivette, MO.
Specifications
Competent Cells E. coli TOP10 | |
20 Reactions |