missing translation for 'onlineSavingsMsg'
Learn More

Abritech Colour filter unmounted for rayboxes 75mm x 50mm

Brand:  Abritech XGB-390-K

Colour filter unmounted for rayboxes 75mm x 50mm

Code : 07

Additional Details : Weight : 0.00376kg

Product Code. 13176230

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.